siRNA Details
| Gene Symbol: RTN4RL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178568.3 | NP_848663.1 | 2 | 1162 | ||
| GenBank mRNA | AF532859.1 | - | 1 | 682 | ||
| AL834409.1 | - | 1 | 628 | |||
| AY250219.1 | - | 1 | 682 | |||
| BC128608.1 | - | 2 | 704 | 40121824 | ||

RTN4RL1
Genomic Map