siRNA Details
| Gene Symbol: DNAJC24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181706.4 | NP_859057.4 | 4 | 398 | ||
| XM_017017216.1 | XP_016872705.1 | 4 | 383 | |||
| GenBank mRNA | AK289665.1 | - | 4 | 386 | ||
| AL833128.1 | - | 4 | 388 | |||
| BC036571.1 | - | 5 | 534 | 4793909 | ||
| BC044931.2 | - | 4 | 350 | |||
| BC063804.1 | - | 4 | 341 | 6474523 | ||
| CD685768.1 | - | 4 | 416 | |||

DNAJC24
Genomic Map