siRNA Details
| Gene Symbol: GPR142 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181790.1 | NP_861455.1 | 1 | 35 | ||
| XM_005257305.3 | XP_005257362.1 | 1 | 344 | |||
| GenBank mRNA | AB196530.1 | - | 1 | 35 | ||
| AY288421.1 | - | 1 | 35 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GPR142| Gene Symbol: GPR142 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181790.1 | NP_861455.1 | 1 | 35 | ||
| XM_005257305.3 | XP_005257362.1 | 1 | 344 | |||
| GenBank mRNA | AB196530.1 | - | 1 | 35 | ||
| AY288421.1 | - | 1 | 35 | |||