siRNA Details
| Gene Symbol: ZNF595 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286052.1 | NP_001272981.1 | 3 | 2281 | ||
| NM_001286053.1 | NP_001272982.1 | 2 | 2154 | |||
| NM_001286054.1 | NP_001272983.1 | 5 | 2478 | |||
| NM_182524.3 | NP_872330.1 | 4 | 2377 | |||
| XM_011513398.2 | XP_011511700.1 | 4 | 2996 | |||
| GenBank mRNA | BX537887.1 | - | 4 | 2329 | ||

ZNF595
Genomic Map