siRNA Details
| Gene Symbol: RNF144B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182757.3 | NP_877434.2 | 8 | 2773 | ||
| XM_005248986.3 | XP_005249043.1 | 7 | 2689 | |||
| GenBank mRNA | AB076367.1 | - | 8 | 2757 | ||
| AL832329.1 | - | 8 | 2598 | |||
| BC063311.1 | - | 8 | 2644 | 30331343 | ||

RNF144B
Genomic Map