siRNA Details
| Gene Symbol: ZNF600 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321866.1 | NP_001308795.1 | 6 | 1769 | ||
| NM_001321867.1 | NP_001308796.1 | 4 | 1146 | |||
| NM_198457.3 | NP_940859.3 | 3 | 1019 | |||
| XM_017026390.1 | XP_016881879.1 | 7 | 1696 | |||
| XM_017026391.1 | XP_016881880.1 | 6 | 1672 | |||
| XM_017026392.1 | XP_016881881.1 | 6 | 1470 | |||
| GenBank mRNA | AK075230.1 | - | 1 | 306 | ||
| AK131231.1 | - | 3 | 1009 | |||
| BX640933.1 | - | 3 | 991 | |||

ZNF600
Genomic Map