siRNA Details
| Gene Symbol: NHLRC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198514.3 | NP_940916.2 | 4 | 1095 | ||
| XM_011539769.2 | XP_011538071.1 | 4 | 1095 | |||
| GenBank mRNA | AK090631.1 | - | 4 | 1066 | ||
| AK098487.1 | - | 2 | 209 | |||
| BX647641.1 | - | 2 | 466 | |||

NHLRC2
Genomic Map