siRNA Details
| Gene Symbol: MAGEA10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011543.2 | NP_001011543.2 | 5 | 1583 | ||
| NM_001251828.1 | NP_001238757.1 | 5 | 1511 | |||
| NM_021048.4 | NP_066386.2 | 4 | 1492 | |||
| GenBank mRNA | BC004105.2 | - | 4 | 1467 | 3926146 | |

MAGEA10
Genomic Map