siRNA Details
| Gene Symbol: SLC25A35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201520.2 | NP_958928.1 | 6 | 1533 | ||
| GenBank mRNA | AK097536.1 | - | 6 | 1042 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC25A35| Gene Symbol: SLC25A35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201520.2 | NP_958928.1 | 6 | 1533 | ||
| GenBank mRNA | AK097536.1 | - | 6 | 1042 | ||