siRNA Details
| Gene Symbol: HIST1H2AD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021065.3 | NP_066409.1 | 1 | 480 | ||
| GenBank mRNA | BC128035.1 | - | 1 | 430 | 40021702 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

HIST1H2AD| Gene Symbol: HIST1H2AD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021065.3 | NP_066409.1 | 1 | 480 | ||
| GenBank mRNA | BC128035.1 | - | 1 | 430 | 40021702 | |