siRNA Details
| Gene Symbol: NPSR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300935.1 | NP_001287864.1 | 3 | 474 | ||
| NM_207172.1 | NP_997055.1 | 3 | 428 | |||
| NM_207173.1 | NP_997056.1 | 3 | 428 | |||
| GenBank mRNA | AK172847.1 | - | 3 | 474 | ||
| AY310326.1 | - | 3 | 428 | |||
| AY310327.1 | - | 3 | 428 | |||
| AY310331.1 | - | 3 | 428 | |||
| AY956446.1 | - | 3 | 333 | |||
| AY956447.1 | - | 3 | 304 | |||
| AY956448.1 | - | 3 | 304 | |||
| BC132693.1 | - | 3 | 345 | 40146715 | ||
| BC132695.1 | - | 3 | 345 | 40146717 | ||
More Information
Set Membership: |
|

NPSR1
Genomic Map