siRNA Details
| Gene Symbol: FSD1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145313.2 | NP_001138785.1 | 8 | 925 | ||
| NM_001287191.1 | NP_001274120.1 | 7 | 829 | |||
| NM_001287192.1 | NP_001274121.1 | 7 | 829 | |||
| XM_005252254.2 | XP_005252311.1 | 8 | 1158 | |||
| XM_005252257.3 | XP_005252314.1 | 7 | 824 | |||
| XM_005252260.3 | XP_005252317.1 | 6 | 791 | |||
| XM_011519077.2 | XP_011517379.1 | 9 | 816 | |||
| XM_011519078.2 | XP_011517380.2 | 8 | 823 | |||
| XM_011519079.2 | XP_011517381.2 | 8 | 825 | |||
| XM_011519080.2 | XP_011517382.1 | 2 | 1521 | |||
| XM_011519081.2 | XP_011517383.1 | 2 | 1506 | |||
| XM_017015182.1 | XP_016870671.1 | 8 | 1158 | |||
| XM_017015183.1 | XP_016870672.1 | 7 | 824 | |||
| XM_017015184.1 | XP_016870673.1 | 3 | 334 | |||
| XM_017015185.1 | XP_016870674.1 | 8 | 831 | |||
| XM_017015186.1 | XP_016870675.1 | 2 | 174 | |||
| XM_017015187.1 | XP_016870676.1 | 7 | 824 | |||
| GenBank mRNA | AK296433.1 | - | 7 | 740 | ||
| AK299350.1 | - | 8 | 857 | |||
| AK299491.1 | - | 7 | 740 | |||
| AK309817.1 | - | 8 | 1166 | |||
| BC036746.1 | - | 2 | 268 | 4384472 | ||

FSD1L
Genomic Map