siRNA Details
| Gene Symbol: FOXN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_213596.2 | NP_998761.2 | 10 | 2578 | ||
| XM_011537922.2 | XP_011536224.1 | 10 | 2767 | |||
| XM_017018818.1 | XP_016874307.1 | 8 | 3553 | |||
| GenBank mRNA | AK093286.1 | - | 3 | 820 | ||
| AK131519.1 | - | 6 | 2185 | |||
| BC146825.1 | - | 10 | 2579 | 8860453 | ||

FOXN4
Genomic Map