siRNA Details
Gene Symbol: PI4KAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC139841.1 | - | 13, 14 | 1574 | ||
CR456378.1 | - | 13, 14 | 1592 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: PI4KAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC139841.1 | - | 13, 14 | 1574 | ||
CR456378.1 | - | 13, 14 | 1592 |