siRNA Details
| Gene Symbol: KRTAP10-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198696.2 | NP_941969.2 | 1 | 61 | ||
| GenBank mRNA | AB076350.1 | - | 1 | 61 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRTAP10-3| Gene Symbol: KRTAP10-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198696.2 | NP_941969.2 | 1 | 61 | ||
| GenBank mRNA | AB076350.1 | - | 1 | 61 | ||