siRNA Details
| Gene Symbol: FAHD2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001320848.1 | NP_001307777.1 | 3 | 504 | ||
| NM_001320849.1 | NP_001307778.1 | 2 | 378 | |||
| NM_199336.2 | NP_955368.1 | 2 | 378 | |||
| XM_011510745.2 | XP_011509047.1 | 3 | 494 | |||
| XM_011510746.2 | XP_011509048.1 | 3 | 613 | |||
| XM_011510747.2 | XP_011509049.1 | 2 | 378 | |||
| XM_011510748.2 | XP_011509050.1 | 3 | 494 | |||
| XM_011510750.2 | XP_011509052.1 | 3 | 496 | |||
| XM_017003470.1 | XP_016858959.1 | 3 | 491 | |||
| XM_017003471.1 | XP_016858960.1 | 3 | 494 | |||
| XM_017003472.1 | XP_016858961.1 | 2 | 370 | |||
| XM_017003473.1 | XP_016858962.1 | 2 | 378 | |||
| GenBank mRNA | AK295056.1 | - | 2 | 331 | ||
| AL833869.1 | - | 1 | 178 | |||
| BC064511.1 | - | 2 | 341 | 6644987 | ||
| BI547630.1 | - | 2 | 383 | |||

FAHD2B
Genomic Map