siRNA Details
| Gene Symbol: CCDC181 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300968.1 | NP_001287897.1 | 4 | 735 | ||
| NM_001300969.1 | NP_001287898.1 | 3 | 437 | |||
| NM_021179.2 | NP_067002.1 | 3 | 437 | |||
| XM_005245383.1 | XP_005245440.1 | 4 | 735 | |||
| XM_011509827.2 | XP_011508129.1 | 3 | 419 | |||
| XM_017001938.1 | XP_016857427.1 | 5 | 2368 | |||
| XM_017001939.1 | XP_016857428.1 | 3 | 400 | |||
| XM_017001940.1 | XP_016857429.1 | 3 | 419 | |||
| GenBank mRNA | AK098712.1 | - | 3 | 380 | ||
| AK223354.1 | - | 3 | 380 | |||
| AK313598.1 | - | 4 | 735 | |||
| AL049687.1 | - | 3 | 395 | |||
| BC026073.2 | - | 3 | 437 | 4827122 | ||

CCDC181
Genomic Map