siRNA Details
| Gene Symbol: EPHB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004444.4 | NP_004435.3 | 4 | 1072 | ||
| XM_017011816.1 | XP_016867305.1 | 4 | 675 | |||
| GenBank mRNA | AB209644.1 | - | 3 | 1782 | ||
| AY056048.1 | - | 4 | 666 | |||
| BC004264.1 | - | 1 | 157 | |||
| BC052804.1 | - | 4 | 749 | 6584384 | ||
| EU826608.1 | - | 4 | 581 | |||
| EU826609.1 | - | 4 | 581 | |||
| EU826610.1 | - | 4 | 581 | |||
| U07695.1 | - | 4 | 666 | |||

EPHB4
Genomic Map