siRNA Details
| Gene Symbol: RNPEPL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018226.4 | NP_060696.4 | 7 | 1309 | ||
| XM_005247036.4 | XP_005247093.1 | 7 | 1370 | |||
| GenBank mRNA | AF300795.1 | - | 7 | 1015 | ||
| AK001668.1 | - | 3 | 273 | |||
| AK130805.1 | - | 2 | 960 | |||
| BC017301.2 | - | 4 | 461 | |||
| BC067258.1 | - | 7 | 884 | 6183046 | ||
| BC082975.1 | - | 7 | 1209 | 6379537 | ||
| BC099906.1 | - | 5 | 1422 | 5311883 | ||

RNPEPL1
Genomic Map