siRNA Details
Gene Symbol: ZP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021186.4 | NP_067009.1 | 1 | 367 | ||
GenBank mRNA | AK314151.1 | - | 1 | 367 | ||
BC069521.1 | - | 1 | 80 | 7262334 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ZP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021186.4 | NP_067009.1 | 1 | 367 | ||
GenBank mRNA | AK314151.1 | - | 1 | 367 | ||
BC069521.1 | - | 1 | 80 | 7262334 |