siRNA Details
| Gene Symbol: WFDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282466.1 | NP_001269395.1 | 3, 4 | 682 | ||
| NM_021197.3 | NP_067020.2 | 3, 4 | 682 | |||
| GenBank mRNA | AF169631.1 | - | 3, 4 | 570 | ||
| AF302109.1 | - | 3, 4 | 593 | |||
| AK075061.1 | - | 3, 4 | 542 | |||
| BC029159.1 | - | 3, 4 | 488 | 5186156 | ||
| CD519223.1 | - | 3, 4 | 596 | |||

WFDC1
Genomic Map