siRNA Details
| Gene Symbol: RPRD1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021215.3 | NP_067038.1 | 2 | 625 | ||
| XM_005260477.3 | XP_005260534.1 | 2 | 709 | |||
| XM_005260480.4 | XP_005260537.1 | 2 | 709 | |||
| XM_011528954.2 | XP_011527256.1 | 2 | 709 | |||
| XM_017027990.1 | XP_016883479.1 | 2 | 709 | |||
| XM_017027991.1 | XP_016883480.1 | 2 | 709 | |||
| GenBank mRNA | AK312468.1 | - | 2 | 385 | ||
| BC001696.2 | - | 2 | 375 | |||
| BC033629.1 | - | 2 | 409 | 5223673 | ||
| DQ372938.1 | - | 2 | 498 | |||

RPRD1B
Genomic Map