siRNA Details
| Gene Symbol: TRIM39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021253.3 | NP_067076.2 | 3 | 431 | ||
| NM_172016.2 | NP_742013.1 | 3 | 745 | |||
| GenBank mRNA | AB046381.1 | - | 2 | 286 | ||
| AI159923.1 | - | 3 | 431 | |||
| AK292512.1 | - | 3 | 348 | |||
| AX776007.1 | - | 3 | 311 | |||
| BC007661.2 | - | 3 | 302 | 3051389 | ||
| BC034985.1 | - | 3 | 718 | 5268723 | ||
| BT007370.1 | - | 1 | 119 | |||
| Gene Symbol: TRIM39-RPP21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199119.1 | NP_001186048.1 | 1 | 126 | ||

TRIM39
Genomic Map