siRNA Details
| Gene Symbol: CDH19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271028.1 | NP_001257957.1 | 3 | 594 | ||
| NM_021153.3 | NP_066976.1 | 3 | 594 | |||
| NR_073130.1 | 3 | 594 | ||||
| XM_011525931.2 | XP_011524233.1 | 3 | 697 | |||
| XM_011525932.1 | XP_011524234.1 | 2 | 372 | |||
| XM_017025711.1 | XP_016881200.1 | 3 | 619 | |||
| GenBank mRNA | AJ007607.1 | - | 3 | 435 | ||
| AK304000.1 | - | 3 | 544 | |||
| AK315257.1 | - | 3 | 596 | |||
| AY358654.1 | - | 2 | 333 | |||
| BC015877.1 | - | 3 | 486 | 4716394 | ||

CDH19
Genomic Map