siRNA Details
| Gene Symbol: WFDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282466.1 | NP_001269395.1 | 5, 6 | 881 | ||
| NM_001282467.1 | NP_001269396.1 | 5, 6 | 878 | |||
| NM_021197.3 | NP_067020.2 | 5, 6 | 881 | |||
| XM_005256079.3 | XP_005256136.1 | 4, 5 | 792 | |||
| GenBank mRNA | AF169631.1 | - | 5, 6 | 769 | ||
| AF302109.1 | - | 5, 6 | 794 | |||
| AK075061.1 | - | 5, 6 | 741 | |||
| AL713785.1 | - | 2, 3 | 3543 | |||
| BC029159.1 | - | 5, 6 | 687 | 5186156 | ||
| BQ670532.1 | - | 5, 6 | 501 | |||
| BQ671964.1 | - | 5, 6 | 501 | |||

WFDC1
Genomic Map