siRNA Details
| Gene Symbol: ZNF71 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021216.4 | NP_067039.1 | 3 | 563 | ||
| XM_011527193.2 | XP_011525495.1 | 4 | 715 | |||
| XM_017027078.1 | XP_016882567.1 | 4 | 625 | |||
| XM_017027079.1 | XP_016882568.1 | 4 | 2495 | |||
| XM_017027080.1 | XP_016882569.1 | 3 | 498 | |||
| GenBank mRNA | AF269249.1 | - | 2 | 409 | ||
| BC014280.2 | - | 3 | 535 | 3840512 | ||

ZNF71
Genomic Map