siRNA Details
| Gene Symbol: NTN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021229.3 | NP_067052.2 | 2 | 787 | ||
| GenBank mRNA | AF119916.3 | - | 3 | 324 | ||
| AF278532.1 | - | 2 | 789 | |||
| AF297711.1 | - | 2 | 639 | |||
| AK226070.1 | - | 2 | 644 | |||
| AK290591.1 | - | 2 | 324 | |||
| AK315796.1 | - | 2 | 646 | |||
| AL833767.1 | - | 1 | 191 | |||
| AY330211.1 | - | 2 | 338 | |||
| BC136798.1 | - | 2 | 762 | 9020788 | ||
| BC136799.1 | - | 2 | 762 | 9020789 | ||
| Z50158.1 | - | 1 | 157 | |||

NTN4
Genomic Map