siRNA Details
| Gene Symbol: RRAGD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021244.4 | NP_067067.1 | 2 | 649 | ||
| XM_005248755.4 | XP_005248812.1 | 2 | 843 | |||
| XM_011536022.2 | XP_011534324.1 | 2 | 421 | |||
| GenBank mRNA | AF272036.1 | - | 2 | 432 | ||
| AK289799.1 | - | 2 | 574 | |||
| BC003088.1 | - | 2 | 311 | 3536088 | ||

RRAGD
Genomic Map