siRNA Details
| Gene Symbol: ANGPTL7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021146.3 | NP_066969.1 | 5 | 2168 | ||
| GenBank mRNA | AB593026.1 | - | 5 | 2102 | ||
| AB593027.1 | - | 5 | 2169 | |||
| AY358301.1 | - | 5 | 2091 | |||
| BC001881.1 | - | 5 | 2116 | 3544149 | ||
| Y16132.1 | - | 5 | 1885 | |||

ANGPTL7
Genomic Map