siRNA Details
Gene Symbol: CD209 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144893.1 | NP_001138365.1 | 5 | 824 | ||
NM_001144894.1 | NP_001138366.1 | 5 | 1100 | |||
NM_001144895.1 | NP_001138367.1 | 7 | 956 | |||
NM_001144896.1 | NP_001138368.1 | 6 | 1160 | |||
NM_001144897.1 | NP_001138369.1 | 7 | 1214 | |||
NM_001144899.1 | NP_001138371.1 | 7 | 749 | |||
NM_021155.3 | NP_066978.1 | 7 | 1232 | |||
NR_026692.1 | 6 | 1332 | ||||
XM_005272472.4 | XP_005272529.1 | 7 | 849 | |||
GenBank mRNA | AF290886.1 | - | 7 | 1174 | ||
AK293089.1 | - | 7 | 1188 | |||
AK304190.1 | - | 7 | 735 | |||
AK308595.1 | - | 4 | 775 | |||
AK313585.1 | - | 7 | 1233 | |||
AY042221.1 | - | 7 | 1165 | |||
AY042222.1 | - | 7 | 1147 | |||
AY042223.1 | - | 7 | 889 | |||
AY042224.1 | - | 6 | 457 | |||
AY042225.1 | - | 6 | 1093 | |||
AY042226.1 | - | 5 | 1033 | |||
AY042227.1 | - | 6 | 757 | |||
AY042229.1 | - | 6 | 1265 | |||
AY042230.1 | - | 5 | 1193 | |||
AY042231.1 | - | 5 | 1055 | |||
BC110615.2 | - | 5 | 1200 | 40032459 | ||
M98457.1 | - | 7 | 1206 |
