siRNA Details
| Gene Symbol: BRINP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021165.3 | NP_066988.1 | 3 | 828 | ||
| XM_005245379.1 | XP_005245436.1 | 4 | 1408 | |||
| GenBank mRNA | AB051534.1 | - | 3 | 821 | ||
| AB161694.1 | - | 3 | 821 | |||
| AK296524.1 | - | 3 | 797 | |||
| AL035289.1 | - | 2 | 402 | |||
| BC028036.1 | - | 3 | 720 | 5242899 | ||

BRINP2
Genomic Map