siRNA Details
| Gene Symbol: SLAMF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282588.1 | NP_001269517.1 | 4 | 1068 | ||
| NM_001282589.1 | NP_001269518.1 | 5 | 1172 | |||
| NM_001282590.1 | NP_001269519.1 | 6 | 1244 | |||
| NM_001282591.1 | NP_001269520.1 | 5 | 1124 | |||
| NM_001282592.1 | NP_001269521.1 | 6 | 1461 | |||
| NM_001282593.1 | NP_001269522.1 | 4 | 1020 | |||
| NM_001282594.1 | NP_001269523.1 | 6 | 1283 | |||
| NM_001282595.1 | NP_001269524.1 | 6 | 1026 | |||
| NM_001282596.1 | NP_001269525.1 | 5 | 1140 | |||
| NM_021181.4 | NP_067004.3 | 7 | 1565 | |||
| XM_011509828.1 | XP_011508130.1 | 7 | 1414 | |||
| XM_011509829.1 | XP_011508131.1 | 6 | 1310 | |||
| GenBank mRNA | AB027233.1 | - | 7 | 1335 | ||
| AK292148.1 | - | 7 | 1349 | |||
| AK298499.1 | - | 5 | 979 | |||
| AK298548.1 | - | 4 | 876 | |||
| AK301137.1 | - | 4 | 811 | |||
| AK301432.1 | - | 6 | 1069 | |||
| AK301438.1 | - | 5 | 913 | |||
| AL713801.1 | - | 2 | 2190 | |||
| AL834424.1 | - | 6 | 1033 | |||
| BC027867.1 | - | 6 | 1227 | 5203638 | ||

SLAMF7
Genomic Map