siRNA Details
| Gene Symbol: ZNF410 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242924.1 | NP_001229853.1 | 14 | 2038 | ||
| NM_001242926.1 | NP_001229855.1 | 11 | 1728 | |||
| NM_001242927.1 | NP_001229856.1 | 11 | 1650 | |||
| NM_001242928.1 | NP_001229857.1 | 13 | 1962 | |||
| NM_021188.2 | NP_067011.1 | 12 | 1869 | |||
| NR_040251.1 | 12 | 1837 | ||||
| GenBank mRNA | AK223261.1 | - | 12 | 1625 | ||
| AK293350.1 | - | 14 | 1794 | |||
| AK298307.1 | - | 13 | 1592 | |||
| AK298520.1 | - | 12 | 1645 | |||
| AK299138.1 | - | 11 | 1648 | |||
| AK303834.1 | - | 11 | 1329 | |||
| AK304609.1 | - | 12 | 1383 | |||
| AK309957.1 | - | 14 | 1783 | |||
| AK315206.1 | - | 12 | 1627 | |||
| BC004357.1 | - | 13 | 1706 | |||
| BC050683.1 | - | 11 | 1538 | 5752452 | ||
| U90919.1 | - | 12 | 1601 | |||

ZNF410
Genomic Map