siRNA Details
| Gene Symbol: RHOU | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021205.5 | NP_067028.1 | 3 | 3867 | ||
| NR_037962.1 | 3 | 3325 | ||||
| GenBank mRNA | AB209511.1 | - | 4 | 3399 | ||
| AF211836.1 | - | 3 | 3823 | |||
| AK001478.1 | - | 1 | 1923 | |||
| BC040076.1 | - | 3 | 3448 | 5768957 | ||

RHOU
Genomic Map