siRNA Details
| Gene Symbol: TRAPPC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166621.1 | NP_001160093.1 | 2, 3 | 164 | ||
| NM_021210.4 | NP_067033.1 | 1, 2 | 224 | |||
| GenBank mRNA | AK314038.1 | - | 1, 2 | 230 | ||
| BC032717.1 | - | 1, 2 | 226 | 5518603 | ||
| BI457771.1 | - | 1, 2 | 226 | |||
| BI600633.1 | - | 2, 3 | 169 | |||
| BM762006.1 | - | 2, 3 | 138 | |||

TRAPPC1
Genomic Map