siRNA Details
| Gene Symbol: NGB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021257.3 | NP_067080.1 | 2, 3 | 580 | ||
| GenBank mRNA | AF422797.1 | - | 2, 3 | 580 | ||
| AJ245946.1 | - | 2, 3 | 205 | |||
| AK098350.1 | - | 2, 3 | 182 | |||
| AK315395.1 | - | 2, 3 | 293 | |||
| BC032509.1 | - | 2, 3 | 287 | 5247233 | ||

NGB
Genomic Map