siRNA Details
| Gene Symbol: PRDM12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021619.2 | NP_067632.2 | 1 | 146 | ||
| GenBank mRNA | AY004252.2 | - | 1 | 146 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PRDM12| Gene Symbol: PRDM12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021619.2 | NP_067632.2 | 1 | 146 | ||
| GenBank mRNA | AY004252.2 | - | 1 | 146 | ||