siRNA Details
| Gene Symbol: C12orf10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021640.3 | NP_067653.3 | 2, 3 | 389 | ||
| GenBank mRNA | AF289485.1 | - | 2, 3 | 357 | ||
| AK293449.1 | - | 2, 3 | 345 | |||
| BC013956.2 | - | 2, 3 | 329 | |||
| BC028904.1 | - | 1, 2 | 293 | 4454943 | ||
| BC051871.1 | - | 2, 3 | 348 | 5170816 | ||

C12orf10
Genomic Map