siRNA Details
| Gene Symbol: ATG101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001098673.1 | NP_001092143.1 | 3 | 415 | ||
| NM_021934.4 | NP_068753.2 | 3 | 586 | |||
| GenBank mRNA | AF218031.1 | - | 3 | 527 | ||
| AK021835.1 | - | 3 | 584 | |||
| BC005151.2 | - | 3 | 538 | 3508173 | ||
| BC009937.2 | - | 3 | 520 | 4303452 | ||
| BP309508.1 | - | 3 | 386 | |||

ATG101
Genomic Map