siRNA Details
| Gene Symbol: IL22RA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021258.3 | NP_067081.2 | 7 | 1363 | ||
| XM_011541882.1 | XP_011540184.1 | 8 | 1312 | |||
| GenBank mRNA | AF286095.1 | - | 7 | 1183 | ||
| AK292204.1 | - | 7 | 1258 | |||
| AK304454.1 | - | 5 | 1108 | |||
| AK308522.1 | - | 8 | 1261 | |||
| AK313971.1 | - | 7 | 1363 | |||
| BC029273.1 | - | 7 | 1199 | 4347189 | ||

IL22RA1
Genomic Map