siRNA Details
| Gene Symbol: RAI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172732.1 | NP_001166203.1 | 3 | 1013 | ||
| NM_001172739.1 | NP_001166210.1 | 3 | 1357 | |||
| NM_001172743.1 | NP_001166214.1 | 3 | 1360 | |||
| NM_021785.4 | NP_068557.3 | 2 | 1163 | |||
| NR_033348.1 | 3 | 1436 | ||||
| NR_033349.1 | 2 | 1242 | ||||
| XM_006724459.2 | XP_006724522.1 | 4 | 1512 | |||
| XM_006724460.1 | XP_006724523.1 | 3 | 1260 | |||
| XM_011545439.2 | XP_011543741.1 | 4 | 1541 | |||
| XM_011545440.2 | XP_011543742.1 | 4 | 1544 | |||
| XM_011545441.2 | XP_011543743.1 | 3 | 1347 | |||
| GenBank mRNA | AK056214.1 | - | 3 | 1256 | ||
| AK298873.1 | - | 3 | 974 | |||
| AK314631.1 | - | 2 | 1126 | |||
| BC027937.1 | - | 3 | 1316 | 5201821 | ||

RAI2
Genomic Map