siRNA Details
| Gene Symbol: CCDC81 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001156474.1 | NP_001149946.1 | 8 | 1384 | ||
| NM_021827.4 | NP_068599.3 | 7 | 1114 | |||
| XM_005274161.1 | XP_005274218.1 | 8 | 1384 | |||
| XM_005274162.3 | XP_005274219.1 | 3 | 285 | |||
| XM_017018105.1 | XP_016873594.1 | 6 | 738 | |||
| GenBank mRNA | AK027167.1 | - | 4 | 556 | ||
| AK131331.1 | - | 7 | 1117 | |||
| AK223168.1 | - | 4 | 562 | |||
| AK298875.1 | - | 4 | 433 | |||
| BC126412.1 | - | 8 | 1228 | 8992128 | ||

CCDC81
Genomic Map