siRNA Details
| Gene Symbol: LRRC17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031692.2 | NP_001026862.1 | 2 | 835 | ||
| NM_005824.2 | NP_005815.2 | 2 | 835 | |||
| XM_005250108.1 | XP_005250165.1 | 3 | 1251 | |||
| GenBank mRNA | AK225701.1 | - | 2 | 732 | ||
| AK226021.1 | - | 2 | 741 | |||
| AK290305.1 | - | 2 | 726 | |||
| AY358889.1 | - | 2 | 671 | |||
| BC027903.1 | - | 2 | 713 | 5229061 | ||
| U32907.1 | - | 2 | 727 | |||

LRRC17
Genomic Map