siRNA Details
| Gene Symbol: PELI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021255.2 | NP_067078.1 | 6 | 5369 | ||
| XM_006720211.3 | XP_006720274.1 | 5 | 7504 | |||
| XM_011536990.2 | XP_011535292.1 | 7 | 5079 | |||
| XM_011536992.2 | XP_011535294.1 | 6 | 5132 | |||
| XM_017021478.1 | XP_016876967.1 | 6 | 5761 | |||
| XM_017021479.1 | XP_016876968.1 | 7 | 14311 | |||
| GenBank mRNA | AF302502.1 | - | 6 | 5263 | ||
| AK025208.1 | - | 1 | 1504 | |||
| AL133605.1 | - | 1 | 3974 | |||

PELI2
Genomic Map