siRNA Details
| Gene Symbol: CHST6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021615.4 | NP_067628.1 | 3 | 2937 | ||
| XM_005255955.4 | XP_005256012.1 | 4 | 3484 | |||
| XM_011523085.2 | XP_011521387.1 | 3 | 3552 | |||
| GenBank mRNA | AF280086.1 | - | 3 | 2945 | ||

CHST6
Genomic Map