siRNA Details
| Gene Symbol: PLAC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001316887.1 | NP_001303816.1 | 4 | 738 | ||
| NM_001316888.1 | NP_001303817.1 | 3 | 766 | |||
| NM_001316889.1 | NP_001303818.1 | 2 | 648 | |||
| NM_021796.3 | NP_068568.1 | 3 | 720 | |||
| XM_011531257.2 | XP_011529559.1 | 3 | 646 | |||
| XM_011531260.2 | XP_011529562.1 | 3 | 760 | |||
| XM_017029236.1 | XP_016884725.1 | 2 | 688 | |||
| GenBank mRNA | AF234654.1 | - | 3 | 725 | ||
| AK075086.1 | - | 3 | 720 | |||
| BC022335.1 | - | 3 | 705 | 4769552 | ||
| BC066327.1 | - | 2 | 648 | 30339131 | ||

PLAC1
Genomic Map