siRNA Details
| Gene Symbol: SCAND2P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003654.2 | 1 | 538 | |||
| NR_004859.1 | 1 | 538 | ||||
| GenBank mRNA | AF229246.1 | - | 1 | 553 | ||
| AK022844.1 | - | 1 | 538 | |||
| AK290489.1 | - | 1 | 548 | |||
| BC011547.1 | - | 1 | 520 | |||
| BC012929.1 | - | 1 | 520 | |||
| BC063836.1 | - | 1 | 494 | |||
| BC093922.1 | - | 1 | 171 | |||
| BC093924.1 | - | 1 | 171 | |||
| BC143241.1 | - | 1 | 171 | |||
| BC143243.1 | - | 1 | 171 | 9051744 | ||

SCAND2P
Genomic Map