siRNA Details
| Gene Symbol: NXF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022052.1 | NP_071335.1 | 2 | 162 | ||
| GenBank mRNA | AF346619.1 | - | 2 | 162 | ||
| AJ277527.1 | - | 2 | 162 | |||
| AK302447.1 | - | 2 | 163 | |||
| BC031616.1 | - | 2 | 155 | 5170537 | ||

NXF3
Genomic Map