siRNA Details
| Gene Symbol: SH2D4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001174159.1 | NP_001167630.1 | 2 | 463 | ||
| NM_022071.3 | NP_071354.2 | 2 | 613 | |||
| XM_011544610.1 | XP_011542912.1 | 2 | 740 | |||
| XM_017013729.1 | XP_016869218.1 | 2 | 462 | |||
| GenBank mRNA | AK024799.1 | - | 2 | 491 | ||
| BC082982.1 | - | 2 | 463 | 6575224 | ||

SH2D4A
Genomic Map