siRNA Details
| Gene Symbol: TP53AIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001251964.1 | NP_001238893.1 | 2 | 1086 | ||
| GenBank mRNA | AB045832.1 | - | 2 | 614 | ||
| AK310612.1 | - | 2 | 772 | |||
| BC069399.1 | - | 1 | 491 | 7262130 | ||
| BC069426.1 | - | 1 | 491 | 7262142 | ||

TP53AIP1
Genomic Map